Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse EOMES Alexa Fluor 488-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Supplier: R&D Systems IC8889G025
This item is not returnable.
View return policy
Description
EOMES Monoclonal specifically detects EOMES in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
EOMES | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
eomesodermin, eomesodermin (Xenopus laevis) homolog, t box, brain, 2, T-box brain protein 2, T-box brain2, TBR2, TBR-2, TBR2eomesodermin homolog, T-brain-2 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
8320 | |
Mouse | |
Purified |
Flow Cytometry | |
1219A | |
Intracellular Staining by Flow Cytometry 5 uL/10^6 cells | |
O54839 | |
Eomes | |
E. coli-derived recombinant mouse EOMES Met1-Ser126 Accession # O54839 | |
25 Tests | |
Primary | |
Detects mouse EOMES in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction