Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse EOMES Antibody, R&D Systems™

Rabbit Monoclonal Antibody has been used in 3 publications
$214.00 - $526.00
Specifications
Antigen | EOMES |
---|---|
Clone | 1219A |
Dilution | Flow Cytometry 0.25 ug/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
EOMES Monoclonal specifically detects EOMES in Mouse samples. It is validated for Flow Cytometry.Specifications
EOMES | |
Flow Cytometry 0.25 ug/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
eomesodermin, eomesodermin (Xenopus laevis) homolog, t box, brain, 2, T-box brain protein 2, T-box brain2, TBR2, TBR-2, TBR2eomesodermin homolog, T-brain-2 | |
EOMES | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
1219A | |
Flow Cytometry | |
Unconjugated | |
Rabbit | |
Mouse | |
O54839 | |
8320 | |
E. coli-derived recombinant mouse EOMES Met1-Ser126 Accession # O54839 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse EOMES in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title