Learn More
Mouse EpCAM/TROP1 Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Supplier:  R&D Systems FAB8998U100UG
Catalog No. FAB8998U100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
EpCAM/TROP1 Monoclonal specifically detects EpCAM/TROP1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
EpCAM/TROP1
Monoclonal
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Rat
100 μg
Primary
Detects mouse EpCAM/TROP-1.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Flow Cytometry
G8.8R
Flow Cytometry 0.25-1 ug/10^6 cells
17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2, GA733-2EGP34, gp40, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A
TE-71 Thymic epithelial cell line
RUO
4072
Mouse
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only (RUO)