Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse EPCR Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | EPCR |
---|---|
Clone | 432732 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB27491SP
![]() |
R&D Systems
MAB27491SP |
25 μg |
Each for $200.50
|
|
|||||
MAB27491
![]() |
R&D Systems
MAB27491 |
100 μg |
Each for $468.00
|
|
|||||
Description
EPCR Monoclonal specifically detects EPCR in Mouse samples. It is validated for Western Blot.Specifications
EPCR | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q64695 | |
10544 | |
Mouse myeloma cell line NS0-derived recombinant mouse EPCR Leu18-Ser214 Accession # Q64695 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse EPCR in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human EPCR is observed. |
432732 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
APC receptor, CCCA, CCD41, CCD41centrosome-associated protein, CD201, CD201 antigen, centrocyclin, Endothelial cell protein C receptor, endothelial protein C receptor, EPCRMGC23024, PROCR, protein C receptor, endothelial | |
PROCR | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title