Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse EphA1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3034
This item is not returnable.
View return policy
Description
EphA1 Polyclonal specifically detects EphA1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
EphA1 | |
Polyclonal | |
Unconjugated | |
AAH71215 | |
EPHA1 | |
Mouse myeloma cell line NS0-derived recombinant mouse EphA1 Glu27-Glu548 Accession # AAH71215 | |
100 μg | |
Primary | |
Detects mouse EphA1 in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant human EphA1 and less than 5% cross-reactivity with recombinant mouse (rm) EphA4, rmEphA6, and rmEphA7 is observed. | |
Mouse | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
LYOPH | |
Western Blot 0.25 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready | |
EC 2.7.10, EC 2.7.10.1, EPH receptor A1, eph tyrosine kinase 1, EphA1, EPHephrin type-A receptor 1, EPHT, EPHT1erythropoietin-producing hepatoma amplified sequence, Esk, MGC163163, oncogene EPH, soluble EPHA1 variant 1, soluble EPHA1 variant 2, Tyrosine-protein kinase receptor EPH | |
Goat | |
Affinity Purified | |
RUO | |
2041 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction