Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse EphA2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Supplier: R&D Systems AF639SP
This item is not returnable.
View return policy
Description
EphA2 Polyclonal specifically detects EphA2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
EphA2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ARCC2, EC 2.7.10, EC 2.7.10.1, Eck, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, Myk2, Sek2, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK | |
Goat | |
Affinity Purified | |
RUO | |
1969 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
AAA82113 | |
EPHA2 | |
Mouse myeloma cell line NS0-derived recombinant mouse EphA2 Ala22-Ala535 Accession # AAA82113 | |
25 μg | |
Primary | |
Detects mouse EphA2 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) EphA2 is observed, and less than 1% cross-reactivity with rhEphA1, recombinant mouse (rm) EphA3, rmEphA4, rmEphA6, rmEphA7, rmEphA8, and recombinant rat EphA5 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction