Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse EphA2 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF639
This item is not returnable.
View return policy
Description
EphA2 Polyclonal specifically detects EphA2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
EphA2 | |
Polyclonal | |
Biotin | |
AAA82113 | |
EPHA2 | |
Mouse myeloma cell line NS0-derived recombinant mouse EphA2 Ala22-Ala535 Accession # AAA82113 | |
50 μg | |
Primary | |
Detects mouse EphA2 in Western blots. In Western blots, less than 2% cross-reactivity with recombinant human EphA1, recombinant mouse (rm) EphA3, rmEphA4, rmEphA6, rmEphA7, rmEphA8 and recombinant rat EphA5 is observed. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
ARCC2, EC 2.7.10, EC 2.7.10.1, Eck, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, Myk2, Sek2, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK | |
Goat | |
Affinity Purified | |
RUO | |
1969 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction