Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse EphA3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB640
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB640 500 μg
MAB640SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB640 Supplier R&D Systems Supplier No. MAB640
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

EphA3 Monoclonal specifically detects EphA3 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen EphA3
Applications Western Blot
Classification Monoclonal
Clone 99437
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P29319
Gene Alias Cek4, EC 2.7.10, EC 2.7.10.1, EK4, EPH receptor A3, EphA3, EPH-like kinase 4, ephrin type-A receptor 3, ETK, hEK4, HEKETK1eph-like tyrosine kinase 1, human embryo kinase 1, Tyro4, TYRO4 protein tyrosine kinase, Tyrosine-protein kinase receptor ETK1, Tyrosine-protein kinase TYRO4
Gene Symbols EPHA3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse EphA3 Glu21-Val541 (predicted) Accession # P29319
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2042
Test Specificity Detects mouse EphA3 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) EphA1, recombinant mouse (rm) EphA2, A4, A6, A7,A8, recombinant rat (rr) EphA5, rrEphB1, rmEphB2, B3, or B6 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.