Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse EphA4 Fc Chimera Recombinant Protein

Catalog No. PIRP88233
Encompass
Click to view available options
Quantity:
200 μg

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (1334 μL) be added to the vial to prepare a stock solution of 0.15 mg/mL. Concentration is measured by UV.

EPHA4 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EphA4 encodes a protein that binds ephrin-A ligands.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q03137
For Use With (Application) ELISA
Format Lyophilized
Formulation PBS with 0.01% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4
Gene ID (Entrez) 13838
Molecular Weight (g/mol) 110 kDa
Name Mouse EphA4 Fc Chimera
Purification Method SDS-PAGE
Quantity 200 μg
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias 2900005C20Rik; AI385584; Cek8; Cek8 protein; EK8; epa4; EPH receptor A4; Epha4; EPH-like kinase 8; Ephrin type-A receptor 4; Hek8; rb; receptor protein-tyrosine kinase HEK8; RGD1560587; Sek; Sek1; Tyro1; TYRO1 protein tyrosine kinase; tyrosine-protein kinase receptor MPK-3; Tyrosine-protein kinase receptor SEK; tyrosine-protein kinase receptor SEK-1; Tyrosine-protein kinase TYRO1
Common Name EphA4
Gene Symbol EPHA4
Biological Activity Measured by its binding ability in a functional ELISA. Immobilized mouse EPHA5 (at 2 μg/ml (100 μL/well) can bind mouse EFNA4-Fc with a linear range of 1.28-32 ng/mL)
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence Mouse EPHA4, amino acids Met1-Thr547 (Accession # NP_031962.2) with a C-terminal human IgG1 Fc region
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Measured by its binding ability in a functional ELISA. Immobilized mouse EPHA5 (at 2 μg/ml (100 μL/well) can bind mouse EFNA4-Fc with a linear range of 1.28-32 ng/mL)
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade >95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.