Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse EphA6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6071
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6071 500 μg
MAB6071SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB6071 Supplier R&D Systems Supplier No. MAB6071
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

EphA6 Monoclonal specifically detects EphA6 in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.

Specifications

Antigen EphA6
Applications Western Blot, Blockade of Receptor-ligand Interaction
Classification Monoclonal
Clone 81605
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q62413
Gene Alias DKFZp434C1418, EC 2.7.10, EC 2.7.10.1, Ehk2, EHK-2, EPA6, EPH homology kinase 2, EPH receptor A6, ephrin receptor EphA6, ephrin type-A receptor 6, FLJ35246, Hek12, PRO57066
Gene Symbols EPHA6
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse EphA6 Met1-Gln546 Accession # Q62413
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285220
Test Specificity Detects mouse EphA6 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant rat EphB1 is observed and no cross-reactivity with recombinant human EphA1, A5, recombinant mouse EphA2, A3, A4, A7, A8, B2, B3, B4, or B6 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.