Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse EphA6 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB607
This item is not returnable.
View return policy
Description
EphA6 Monoclonal specifically detects EphA6 in Mouse samples. It is validated for Western Blot.Specifications
EphA6 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DKFZp434C1418, EC 2.7.10, EC 2.7.10.1, Ehk2, EHK-2, EPA6, EPH homology kinase 2, EPH receptor A6, ephrin receptor EphA6, ephrin type-A receptor 6, FLJ35246, Hek12, PRO57066 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
285220 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot | |
81610 | |
Western Blot 1 ug/mL | |
Q62413 | |
EPHA6 | |
Mouse myeloma cell line NS0-derived recombinant mouse EphA6 Ser28-Gln546 Accession # Q62413 | |
500 μg | |
Primary | |
Detects mouse EphA6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human EphA1, A5, recombinant mouse EphA2, A3, A4, A7, A8, B2, B3, B4, B6, or recombinant rat EphB1 is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction