Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse EphA8 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | EphA8 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
EphA8 Polyclonal specifically detects EphA8 in Mouse samples. It is validated for Western Blot.Specifications
EphA8 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
EEK, EK3, EPH- and ELK-related kinase, EPH- and ELK-related tyrosine kinase, EPH receptor A8, EPH-like kinase 3, ephrin type-A receptor 8, HEK3, hydroxyaryl-protein kinase, KIAA1459, protein-tyrosine kinase, tyrosine-protein kinase receptor EEK, tyrosylprotein kinase | |
EPHA8 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
O09127 | |
2046 | |
Mouse myeloma cell line NS0-derived recombinant mouse EphA8 Gly27 - Arg540 Accession # O09127 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects recombinant mouse EphA8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant rat EphA5, recombinant mouse (rm) EphA6, rmEphA7, rmEphA2, recombinant human EphA1, and rmEphA3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title