Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse EphB4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF446
This item is not returnable.
View return policy
Description
EphB4 Polyclonal specifically detects EphB4 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
EphB4 | |
Polyclonal | |
Unconjugated | |
P54761 | |
EPHB4 | |
Mouse myeloma cell line NS0-derived recombinant mouse EphB4 Leu16-Ala539 Accession # P54761 | |
100 μg | |
Primary | |
Detects mouse EphB4 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse (rm) EphB6 and rmEphA3 is observed and less than 1% cross-reactivity with rmEphA2, rmEphA4, rmEphA6, rmEphA7, rmEphA8, rmEphB2, and rmEphB3 is observed. | |
Mouse | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready | |
EC 2.7.10, EC 2.7.10.1, EPH receptor B4, EphB4, ephrin type-B receptor 4, hepatoma transmembrane kinase, Htk, HTKephrin receptor EphB4, Mdk2, Myk1, soluble EPHB4 variant 1, soluble EPHB4 variant 2, soluble EPHB4 variant 3, Tyro11, Tyrosine-protein kinase receptor HTK, Tyrosine-protein kinase TYRO11 | |
Goat | |
Affinity Purified | |
RUO | |
2050 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction