Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse EphB6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF611
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF611 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF611 Supplier R&D Systems Supplier No. AF611
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 6 publications

EphB6 Polyclonal specifically detects EphB6 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen EphB6
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. O08644
Gene Alias EC 2.7.10.1, EPH receptor B6, EphB6, ephrin type-B receptor 6, Hep, HEPTyrosine-protein kinase-defective receptor EPH-6, Mep, MGC129910, MGC129911
Gene Symbols EPHB6
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse EphB6 Leu33-Ser587 Accession # O08644
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2051
Test Specificity Detects mouse EphB6 in direct ELISAs and Western blots. In Western blots, less than 1% cross-reactivity with recombinant mouse (rm) EphB2, rmEphB3, and rmEphB4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.