Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse EphB6 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF611
This item is not returnable.
View return policy
Description
EphB6 Polyclonal specifically detects EphB6 in Mouse samples. It is validated for Western Blot.Specifications
EphB6 | |
Polyclonal | |
Biotin | |
O08644 | |
EPHB6 | |
Mouse myeloma cell line NS0-derived recombinant mouse EphB6 Leu33-Ser587 Accession # O08644 | |
50 μg | |
Primary | |
Detects mouse EphB6 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) EphA1, recombinant mouse (rm) EphA3, rmEphA4, rmEphrin-B1, rmEphA6, rmEphA7, recombinant rat EphA5, rmEphrin-B2, rmEphB1, rmEphB2, rmEphB3, rmEphB4, rrEphB1, rmEphA8, rmEphA2, and rhEphrin-B3 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
EC 2.7.10.1, EPH receptor B6, EphB6, ephrin type-B receptor 6, Hep, HEPTyrosine-protein kinase-defective receptor EPH-6, Mep, MGC129910, MGC129911 | |
Goat | |
Affinity Purified | |
RUO | |
2051 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction