Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Ephrin-A1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF702
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF702 100 μg
AF702SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF702 Supplier R&D Systems Supplier No. AF702
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

Ephrin-A1 Polyclonal specifically detects Ephrin-A1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Ephrin-A1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. P52793
Gene Alias B61, ECKLG, EFL1, EFL-1, EFNA1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EphrinA1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4
Gene Symbols EFNA1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Ephrin-A1 Asp19-Ser182 Accession # P52793
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1942
Test Specificity Detects mouse Ephrin-A1 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) Ephrin-A1 is observed and less than 1% cross-reactivity with recombinant mouse (rm) Ephrin-B1, rmEphrin-A5, rmEphrin-A4, rmEphrin-A2, and rmEphrin-B1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.