Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Ephrin-A2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB458
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB458 500 μg
MAB458SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB458 Supplier R&D Systems Supplier No. MAB458
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Ephrin-A2 Monoclonal specifically detects Ephrin-A2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Ephrin-A2
Applications Western Blot
Classification Monoclonal
Clone 84019
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P52801
Gene Alias Cek7-L, EFNA2, ELF-1, EPH-related receptor tyrosine kinase ligand 6, ephrin-A2, EphrinA2, EPLG6HEK7 ligand, HEK7-L, HEK7-ligand, LERK6, LERK6LERK-6, ligand of eph-related kinase 6
Gene Symbols EFNA2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Ephrin-A2 Arg21-Asn184 Accession # P52801
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1943
Test Specificity Detects mouse Ephrin-A2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant mouse (rm) Ephrin-A4, recombinant human (rh) Ephrin-A3, rmEphrin-B1, and rhEphrin-B3 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.