Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Ephrin-A2 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF603
This item is not returnable.
View return policy
Description
Ephrin-A2 Polyclonal specifically detects Ephrin-A2 in Mouse samples. It is validated for Western Blot.Specifications
Ephrin-A2 | |
Polyclonal | |
Biotin | |
P52801 | |
EFNA2 | |
Mouse myeloma cell line NS0-derived recombinant mouse Ephrin-A2 Met1-Asn184 Accession # P52801 | |
50 μg | |
Primary | |
Detects mouse Ephrin-A2 in Western blots. In Western blots, less than 2% cross-reactivity with recombinant human Ephrin-A5 and recombinant mouse Ephrin-A4 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Cek7-L, EFNA2, ELF-1, EPH-related receptor tyrosine kinase ligand 6, ephrin-A2, EphrinA2, EPLG6HEK7 ligand, HEK7-L, HEK7-ligand, LERK6, LERK6LERK-6, ligand of eph-related kinase 6 | |
Goat | |
Affinity Purified | |
RUO | |
1943 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction