Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Ephrin-B1 Biotinylated Antibody, R&D Systems™
GSA_VAGREENER_CHOICE

Catalog No. BAF473
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF473 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF473 Supplier R&D Systems Supplier No. BAF473
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Ephrin-B1 Polyclonal specifically detects Ephrin-B1 in Mouse samples. It is validated for Western Blot.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Antigen Ephrin-B1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. AAA53231
Gene Alias Cek5-L, EFL3, EFL-3, EFNB1, ELK ligand, Elk-L, ephrin-B1, EphrinB1, LERK-2, STRA-1
Gene Symbols Efnb1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Ephrin-B1 Lys30-Ser229 Accession # AAA53231
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1947
Test Specificity Detects mouse Ephrin-B1 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) Ephrin-A5, recombinant mouse (rm) Ephrin-A1, rhEphrin-A3, rmEphrin-A2, rmEphrin-B2, and rmEphrin-B3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.