Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Epigen Antibody, R&D Systems™

Goat Polyclonal Antibody
$188.28 - $544.12
Specifications
| Antigen | Epigen |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
Description
Epigen Polyclonal specifically detects Epigen in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Epigen | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| ALGV3072, EPG, EPGN, PRO9904 | |
| Epgn | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Q924X1.1 | |
| 255324 | |
| E. coli-derived recombinant mouse Epigen Leu53-Ala103 Accession # Q924X1.1 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Epigen in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title