Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Epiregulin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1068SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1068SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1068SP Supplier R&D Systems Supplier No. MAB1068SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Epiregulin Monoclonal specifically detects Epiregulin in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Epiregulin
Applications Neutralization, ELISA
Classification Monoclonal
Clone 189611
Concentration LYOPH
Conjugate Unconjugated
Dilution Neutralization 0.2-1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q61521
Gene Alias epiregulin, ER, EREG, proepiregulin
Gene Symbols EREG
Host Species Rat
Immunogen E. coli-derived recombinant mouse Epiregulin Val56-Leu101 Accession # Q61521
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2069
Test Specificity Detects mouse Epiregulin in ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human Epiregulin is observed, but no cross-reactivity with recombinant mouse (rm) Epigen, recombinant human (rh) TGF-alpha , rhAmphiregulin, rmBetacellulin, rhEGF, rhHB-EGF, rhHRG-alpha , or rhHRG-beta is observed. In sandwich immunoassays, no cross-reactivity with rmAmphiregulin, rhNRG-1-beta 1, rmBetacellulin, rmEGF, rhHeregulin, rhHB-EGF, or rhTGF-alpha is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.