Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Erythropoietin/EPO Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
$200.50 - $2020.00
Specifications
| Antigen | Erythropoietin/EPO |
|---|---|
| Clone | 148438 |
| Dilution | Neutralization 0.2-0.8 ug/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
Description
Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Mouse samples. It is validated for Neutralization.Specifications
| Erythropoietin/EPO | |
| Neutralization 0.2-0.8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 | |
| EPO | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 148438 | |
| Neutralization | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P07321 | |
| 2056 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Erythropoietin/EPO, Ala27-Arg192, Accession # P07321 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Erythropoietin/EPO. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title