Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse Erythropoietin/EPO Quantikine ELISA Kit

Catalog No. mep00b
Click to view available options
Quantity:
1 x 96-well Plate
This item is not returnable. View return policy
This item is not returnable. View return policy

Mouse Erythropoietin Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse Erythropoietin Quantikine ELISA Kit quantifies mouse Erythropoietin in cell culture supernates (50 ul), tissue homogenates (50 ul), serum (25 ul), heparin plasma (25 ul).

Specifications

Assay Range 47.00 - 3000pg/mL
Assay Sensitivity 46.9pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (Heparin), Serum, Tissue Homogenates
Specificity Natural and Recombinant Mouse Epo. Recombinant Rat Epo has 100% cross-reactivity in this kit. This kit is not validated for use with Rat samples, because Rat sample dilution is not linear.
For Use With (Application) ELISA
Gene ID (Entrez) 13856
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Tissue Homogenates (50uL),Serum (25uL),Heparin Plasma (25uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Target Erythropoietin/EPO
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.