Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse ESGP Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF4580SP
This item is not returnable.
View return policy
Description
ESGP Polyclonal specifically detects ESGP in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
ESGP | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Gm7325 | |
E. coli-derived recombinant mouse ESGP Leu24-Lys84 Accession # Q2Q5T5 | |
25 μg | |
Primary | |
Detects mouse ESGP in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Q2Q5T5 | |
Sheep | |
Affinity Purified | |
RUO | |
653016 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction