Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse Estrogen Receptor beta (aa 467-485) Synthetic Peptide

Catalog No. PEP028
Click to view available options
Quantity:
50 μg

Synthetic Peptide

Mouse Estrogen Receptor B (ERb) (residues 467-485) Synthetic Peptide, PEP-028, from Invitrogen.

Estrogen Receptor beta (ER beta, NR3A2) protein is approximately 55kD, though multiple isoforms ranging from 36-59kD have been described. Human ER beta was initially cloned and characterized from testis. Estrogen Receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of nuclear receptors. The estrogen receptor is a ligand-activated transcription factor, that when bound to estrogen hormone, induces a conformational change that allows dimerization and binding to estrogen response elements (ERE) in DNA. When bound to EREs, ER can positively or negatively regulate gene transcription through the recruitment of coactivator or corepressor proteins. There are two different forms of the estrogen receptor, alpha and beta, encoded by separate genes (ESR1 and ESR2, respectively). Estrogen receptor beta (ER beta) binds estrogens with an affinity similar to that of estrogen receptor alpha. Some ER beta isoforms dominantly inhibit the activity of estrogen receptor alpha in reproductive tissues. ER beta is found widely in many tissues throughout the body and can act as a potent tumor suppressor, playing a crucial role in many cancer types such as prostate cancer, autism spectrum disorder, medullary thyroid carcinoma, De Quervain Disease and risk of cardiovascular disease. PEP-028 is a 19 amino acid (amino acids 467-485) synthetic peptide whose sequence corresponds to the C-terminal of mouse ERb. The sequence of this peptide is (amino to carboxy terminus): C467-C-S-T-E-D-S-K-N-K-E-G-S-Q-N-L-Q-S-Q485This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and ERb , catalog &Num; PA1-312. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 mL of distilled water.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 13983
Species Mouse
Purification Method purified
Accession Number O08537
Common Name Mouse Estrogen Receptor beta (aa 467-485)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol Esr2
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence C467 - C - S - T - E - D - S - K - N - K - E - G - S - Q - N - L - Q - S - Q485
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.