Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Exostosin-like 2/EXTL2 Lumenal Domain Antibody, R&D Systems™

Goat Polyclonal Antibody
$176.44 - $520.69
Specifications
| Antigen | Exostosin-like 2/EXTL2 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Exostosin-like 2/EXTL2 Polyclonal specifically detects Exostosin-like 2/EXTL2 in Mouse samples. It is validated for Western Blot.Specifications
| Exostosin-like 2/EXTL2 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| Alpha-1,4-N-acetylhexosaminyltransferase EXTL2, alpha-1,4-N-acteylhexosaminyltransferase, Alpha-GalNAcT EXTL2, EC 2.4.1.223, exostoses (multiple)-like 2, Exostosin like 2, exostosin-like 2, EXTL2, EXTR2, EXT-related protein 2, Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase, processed exostosin-like 2 | |
| EXTL2 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9ES89 | |
| 2135 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Exostosin-like 2/EXTL2 Lumenal Domain Asn43-Met330 Accession # Q9ES89 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Exostosin-like 2/EXTL2 Lumenal Domain in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse EXTL1 and recombinant human EXTL3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title