Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse F4/80 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB5580R100UG
This item is not returnable.
View return policy
Description
F4/80 Monoclonal specifically detects F4/80 in Mouse samples. It is validated for Flow Cytometry.Specifications
F4/80 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q61549 | |
EMR1 | |
NS0 mouse myeloma cell line transfected with mouse F4/80/EMR1 Gln28-Gly931 Accession # Q61549 | |
RUO | |
2015 | |
Mouse | |
IgG2a |
Flow Cytometry | |
521204 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
ADGRE1, Adhesion G protein-coupled receptor E1, cell surface glycoprotein F4/80, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, EMR1 hormone receptor, F4/80, Gpf480, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7LN3 | |
Rat | |
100 μg | |
Primary | |
Detects mouse F4/80/EMR1. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction