Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse FABP9/T-FABP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2750SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2750SP 25 μg
AF2750 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2750SP Supplier R&D Systems Supplier No. AF2750SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

FABP9/T-FABP Polyclonal specifically detects FABP9/T-FABP in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen FABP9/T-FABP
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O08716
Gene Alias FABP-9, Fatty Acid Binding Protein 9, Testis, Fatty Acid-Binding Protein 9, Lipid-Binding Protein, PERF, PERF 15, PERF15, Testis Lipid Binding Protein, Testis Lipid-Binding Protein, Testis-Type Fatty Acid-Binding Protein, TFABP, T-FABP, TLBP
Gene Symbols Fabp9
Host Species Goat
Immunogen E. coli-derived recombinant mouse FABP9/T-FABP Met1-Val132 Accession # O08716
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 646480
Test Specificity Detects mouse FABP9/T-FABP in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human FABP3 and recombiant mouse FABP4 is observed and less than 1% cross-reactivity with recombiant rat (rr) FABP1 and rrFABP2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.