Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse FAM3B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2867
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2867 100 μg
AF2867SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2867 Supplier R&D Systems Supplier No. AF2867
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

FAM3B Polyclonal specifically detects FAM3B in Mouse samples. It is validated for Western Blot.

Specifications

Antigen FAM3B
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9D309
Gene Alias 2-21, C21orf11, C21orf76PANDER, chromosome 21 open reading frame 11, Cytokine-like protein 2-21, D21M16SJHU19e, family with sequence similarity 3, member B, ORF9, pancreatic derived factor, Pancreatic-derived factor, PANDER, PRED44
Gene Symbols FAM3B
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse FAM3B Glu30-Arg235 Accession # Q9D309
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54097
Test Specificity Detects mouse FAM3B in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant human FAM3B is observed, approximately 5% cross-reactivity with recombinant mouse (rm) FAM3C is observed, and less than 1% cross-reactivity with rmFAM3D is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.