Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse FAM3C Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | FAM3C |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
FAM3C Polyclonal specifically detects FAM3C in Mouse samples. It is validated for Western Blot.Specifications
FAM3C | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
D6WSU176e, family with sequence similarity 3, member C, GS3876, ILEI, ILEIInterleukin-like EMT inducer, predicted osteoblast protein | |
FAM3C | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q91VU0 | |
10447 | |
Mouse myeloma cell line NS0-derived recombinant mouse FAM3C Gln25-Asp227 Accession # Q91VU0 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse FAM3C in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant human FAM3C is observed and less than 5% cross-reactivity with recombinant mouse (rm) FAM3B and rmFAM3D is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title