Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Fas Ligand/TNFSF6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB526SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB526SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB526SP Supplier R&D Systems Supplier No. MAB526SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Fas Ligand/TNFSF6 Monoclonal specifically detects Fas Ligand/TNFSF6 in Mouse samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen Fas Ligand/TNFSF6
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 101624
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 10-20 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P41047
Gene Alias apoptosis (APO-1) antigen ligand 1, Apoptosis antigen ligand, APT1LG1CD95L, APTL, CD178, CD178 antigen, CD95L, CD95-L, Fas antigen ligand, Fas ligand, Fas ligand (TNF superfamily, member 6), FASLCD95 ligand, FASLG, TNFSF6, TNFSF6FasL, tumor necrosis factor (ligand) superfamily, member 6, tumor necrosis factor ligand superfamily member 6
Gene Symbols Fasl
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Fas Ligand/TNFSF6 Pro132-Leu279 Accession # P41047
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 356
Test Specificity Detects mouse Fas Ligand/TNFSF6 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) TRAIL and recombinant mouse (rm) TRANCE is observed and 5% cross-reactivity with rhFas Ligand, recombinant rat Fas Ligand, rmTNF-alpha , rhAPRIL, and rhVEGI is observed and no cross-reactivity with rhGITR Ligand and rhLIGHT is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.