Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Fas Ligand/TNFSF6 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF526
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF526 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF526 Supplier R&D Systems Supplier No. BAF526
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Fas Ligand/TNFSF6 Polyclonal specifically detects Fas Ligand/TNFSF6 in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen Fas Ligand/TNFSF6
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. Q544E9
Gene Alias apoptosis (APO-1) antigen ligand 1, Apoptosis antigen ligand, APT1LG1CD95L, APTL, CD178, CD178 antigen, CD95L, CD95-L, Fas antigen ligand, Fas ligand, Fas ligand (TNF superfamily, member 6), FASLCD95 ligand, FASLG, TNFSF6, TNFSF6FasL, tumor necrosis factor (ligand) superfamily, member 6, tumor necrosis factor ligand superfamily member 6
Gene Symbols Fasl
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Fas Ligand/TNFSF6 Pro132-Leu279 Accession # Q544E9
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 356
Test Specificity Detects mouse Fas Ligand/TNFSF6 in ELISAs and Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human Fas Ligand/TNFSF6 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.