Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Fas/TNFRSF6/CD95 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF435SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF435SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF435SP Supplier R&D Systems Supplier No. AF435SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

Fas/TNFRSF6/CD95 Polyclonal specifically detects Fas/TNFRSF6/CD95 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Fas/TNFRSF6/CD95
Applications Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P25446
Gene Alias Apo-1, Apo-1 antigen, apoptosis antigen 1, Apoptosis-mediating surface antigen FAS, APT1, APT1FASTM, CD95, CD95 antigen, CD95ALPS1A, Fas (TNF receptor superfamily, member 6), Fas AMA, Fas antigen, FAS1, FASLG receptor, TNFRSF6, TNFRSF6member 6, tumor necrosis factor receptor superfamily member 6
Gene Symbols FAS
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Fas/TNFRSF6/CD95 Gly14-Arg169 Accession # P25446
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 355
Test Specificity Detects mouse Fas/TNFRSF6/CD95 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human (rh) Fas, rh4-1BB, rhDR6, rhNGF R, recombinant mouse (rm) OPG, rmRANK and rmCD30 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.