Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Fc gamma RI/CD64 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2074
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2074 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2074 Supplier R&D Systems Supplier No. BAF2074
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Fc gamma RI/CD64 Polyclonal specifically detects Fc gamma RI/CD64 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Fc gamma RI/CD64
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. P26151
Gene Alias CD64, CD64 antigen, CD64a, CD64b, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FCGR1B, FcgRI, FcgRIA, FCRI, FcRIA, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I
Gene Symbols Fcgr1
Host Species Goat
Immunogen NS0-derived recombinant mouse Fc gamma RI/CD64 Glu25-Pro297 Accession # P26151
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2209
Test Specificity Detects mouse Fc gamma RI/CD64 in Western blots. In Western blots, approximately 35% cross-reactivity with recombinant human Fc gamma RI is observed and less than 1% cross-reactivity with recombinant mouse (rm) Fc gamma RIIb and rmFc gamma RIII is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.