Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Fc gamma RIIB/CD32b Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. MAB14601SP Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB14601SP 25 μg
MAB14601100 100 μg
MAB14601500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB14601SP Supplier R&D Systems Supplier No. MAB14601SP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

Fc gamma RIIB/CD32b Monoclonal specifically detects Fc gamma RIIB/CD32b in Human∣Mouse samples. It is validated for Co-immunoprecipitation (Co-IP)∣ELISA Capture (Matched Antibody Pair)∣Flow Cytometry∣Functional Assay∣Immunohistochemistry∣Immunohistochemistry (Frozen)∣Western Blot.

Specifications

Antigen Fc gamma RIIB/CD32b
Applications Co-immunoprecipitation (Co-IP), ELISA Capture (Matched Antibody Pair), Flow Cytometry, Functional Assay, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 190907
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P08101
Gene Alias CD32 antigen, CD32b, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, FCGR2B, FcgRIIB, fcRII-b, FCRIIB, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b
Gene Symbols FCGR2B
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Fc gamma RIIB aa 40-217 Accession # P08101
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2213
Test Specificity Detects mouse Fc gamma RIIB in direct ELISAs. In direct ELISAs, this antibody shows 100% cross-reactivity with recombinant mouse (rm) Fc gamma RIII. In ELISAs, this antibody shows approximately 7% cross-reactivity with rmFc gamma RIII.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.