Learn More
Mouse Fc gamma RIII (CD16) Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
Description
Specifications
Specifications
| Antigen | Fc gamma RIII (CD16) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 275003 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Accession No. | Q5D5J5 |
| Gene Alias | CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20 |
| Gene Symbols | Fcgr3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.