Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Fc gamma RIII (CD16) PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB19601P02
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB19601P02 25 Tests
FAB19601P10 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB19601P02 Supplier R&D Systems Supplier No. FAB19601P025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Fc gamma RIII (CD16) Monoclonal specifically detects Fc gamma RIII (CD16) in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Fc gamma RIII (CD16)
Applications Flow Cytometry
Classification Monoclonal
Clone 275003
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q5D5J5
Gene Alias CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20
Gene Symbols Fcgr3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Fc gamma RIII Ala31-Thr215 Accession # Q5D5J5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2214
Test Specificity Detects mouse Fc gamma RIIIA/B (CD16) in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Fcrl3/CD16-2, rmFc gamma RIA or rhFc gamma RIIA is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.