Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse FCRL1/FcRH1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7286
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7286 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7286 Supplier R&D Systems Supplier No. MAB7286
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

FCRL1/FcRH1 Monoclonal specifically detects FCRL1/FcRH1 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen FCRL1/FcRH1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 745916
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8R4Y0
Gene Alias BXMH1, CD307a, CD307a antigen, Fc receptor homolog 1, Fc receptor-like 1, FcRH1, FCRH1DKFZp667O1421, FcRL1, FcR-like protein 1, hIFGP1, IFGP family protein 1, IFGP1, IFGP1Fc receptor-like protein 1, Immune receptor translocation-associated protein 5, IRTA5, IRTA5immunoglobulin superfamily Fc receptor, gp42
Gene Symbols FCRL1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse FCRL1/FcRH1 Ala17-Ser219 Accession # Q8R4Y0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115350
Test Specificity Detects mouse FCRL1/FcRH1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human FCRL1/FcRH1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.