Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse FCRL5/FcRH5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6757
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6757 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6757 Supplier R&D Systems Supplier No. AF6757
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

FCRL5/FcRH5/IRTA2 Polyclonal specifically detects FCRL5/FcRH5/IRTA2 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen FCRL5/FcRH5/IRTA2
Applications Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Gene Accession No. NP_899045
Gene Alias BXMAS1, BXMAS1MGC119592, CD307, CD307e, CD307e antigen, DKFZp667E2019, DKFZp667F216, Fc receptor homolog 5, Fc receptor-like 5, Fc receptor-like protein 5, FcRH5, FCRH5MGC119593, FcRL5, FcR-like protein 5, FLJ00333, FLJ00397, IFGP5, Immune receptor translocation-associated protein 2, immunoglobulin superfamily receptor translocation associated 2 (IRTA2), IRTA2, IRTA2MGC119590, PRO820
Gene Symbols FCRL5
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse FCRL5/FcRH3 Gln27-Ala496 (predicted) Accession # NP_899045
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83416
Test Specificity Detects mouse FCRL5/FcRH3 in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human FCRL5 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.