Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse FCRLA Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB7905
This item is not returnable.
View return policy
Description
FCRLA/FCRLM1 Monoclonal specifically detects FCRLA/FCRLM1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
FCRLA/FCRLM1 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Q920A9 | |
FCRLA | |
E. coli-derived recombinant mouse FCRLA Lys221-Lys352 Accession # Q920A9 | |
100 μg | |
Primary | |
Detects mouse FCRLA in ELISA. | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
865910 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Fc receptor homolog expressed in B cells (FREB), Fc receptor homolog expressed in B-cells, Fc receptor related protein X, Fc receptor-like A, Fc receptor-like and mucin-like 1, Fc receptor-related protein X, FCRL1, FCRLa, FCRLb, FCRLc1, FCRLc2, FCRLd, FCRLe, FCRLFc receptor-like protein, FCRLM1, FCRLX, FCRX, FREB, FREBFc receptor-like and mucin-like protein 1, MGC4595 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
84824 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction