Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse FCRLB/FCRY Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | FCRLB/FCRY |
---|---|
Clone | 465905 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
FCRLB/FCRY Monoclonal specifically detects FCRLB/FCRY in Mouse samples. It is validated for Western Blot.Specifications
FCRLB/FCRY | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q5DRQ8 | |
127943 | |
Mouse myeloma cell line NS0-derived recombinant mouse FCRLB/FCRY Ala18-Ser427 Accession # Q5DRQ8 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse FCRLB/FCRY in direct ELISAs and Western blots. In Western blots, 75% cross-reactivity with recombinant human FCRLB is observed. |
465905 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Fc receptor homolog expressed in B cells protein 2, Fc receptor like 2, Fc receptor-like and mucin-like 2, Fc receptor-like and mucin-like protein 2, Fc receptor-like B, Fc receptor-like protein 2, Fc receptor-related protein Y, FcRL2, FCRL2FCRLM2, FCRLM2, FCRLY, FCRY, FLJ31052, FREB2, FREB-2, FREB-2FcRY, RP11-474I16.6 | |
FCRLB | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title