Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Fetuin A/AHSG Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Fetuin A/AHSG |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Fetuin A/AHSG Polyclonal specifically detects Fetuin A/AHSG in Mouse samples. It is validated for Western Blot.Specifications
Fetuin A/AHSG | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
A2HS, AHS, AHSG, alpha-2-HS-glycoprotein, alpha-2-Z-globulin, Ba-alpha-2-glycoprotein, FETUAba-alpha-2-glycoprotein, fetuin-A, HSGA | |
AHSG | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
NP_038493 | |
197 | |
Mouse myeloma cell line NS0-derived recombinant mouse Fetuin A/AHSG (R&D Systems, Catalog # 1563-PI) Ala19-Ile345 Accession # NP_038493 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Fetuin A/AHSG in direct ELISAs and Western blots. In these formats, approximately 5% cross-reactivity with recombinant human Fetuin A/AHSG is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title