Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Fetuin A/AHSG Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $667.00
Specifications
Antigen | Fetuin A/AHSG |
---|---|
Clone | 207440 |
Concentration | LYOPH |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB15632SP
![]() |
R&D Systems
MAB15632SP |
25 μg |
Each for $200.50
|
|
|||||
MAB15632
![]() |
R&D Systems
MAB15632 |
500 μg |
Each for $667.00
|
|
|||||
Description
Fetuin A/AHSG Monoclonal specifically detects Fetuin A/AHSG in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
Fetuin A/AHSG | |
LYOPH | |
ELISA | |
Unconjugated | |
Rat | |
Mouse | |
P29699 | |
197 | |
Mouse myeloma cell line NS0-derived recombinant mouse Fetuin A/AHSG Ala19-Ile345 Accession # P29699 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Fetuin A/AHSG in ELISAs. In sandwich immunoassays, no cross-reactivty or interference was observed with recombinant human Fetuin A, rhTGF-beta 1, recombinant mouse (rm) BMP-4, rmMMP-2, recombinant rat MMP-2, or rmMMP-9 is observed. |
207440 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
A2HS, AHS, AHSG, alpha-2-HS-glycoprotein, alpha-2-Z-globulin, Ba-alpha-2-glycoprotein, FETUAba-alpha-2-glycoprotein, fetuin-A, HSGA | |
AHSG | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title