Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse FGF-23 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 8 publications
Supplier: R&D Systems MAB26291
This item is not returnable.
View return policy
Description
FGF-23 Monoclonal specifically detects FGF-23 in Mouse samples. It is validated for Immunohistochemistry.Specifications
FGF-23 | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
Q9EPC2 | |
Fgf23 | |
Mouse myeloma cell line NS0-derived recombinant mouse FGF-23 Tyr25-Val251 (Arg179Gln) Accession # Q9EPC2 | |
100 μg | |
Primary | |
Detects mouse FGF-23 in direct ELISAs and Western blots. In direct ELISAs, this antibody shows approximately 50% cross-reactivity with recombinant human (rh) FGF-23. In Western blots, this antibody does not cross-react with rhFGF-3, -4, -5, -7, -9, -10, -11, -12, -13, -16, -17, -18, -19, acidic, basic, rmFGF-8b, -8c, -15, or -21. | |
Mouse | |
Purified |
Immunohistochemistry | |
283507 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ADHR, FGF23, FGF-23, fibroblast growth factor 23, HPDR2, HYPF, Phosphatonin, PHPTC, tumor-derived hypophosphatemia inducing factor, Tumor-derived hypophosphatemia-inducing factor | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8074 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction