Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse FGF-4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5846
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5846 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5846 Supplier R&D Systems Supplier No. MAB5846
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

FGF-4 Monoclonal specifically detects FGF-4 in Mouse samples. It is validated for Immunohistochemistry.

Specifications

Antigen FGF-4
Applications Immunohistochemistry
Classification Monoclonal
Clone 678431
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_034332
Gene Alias FGF4, fibroblast growth factor 4, HBGF-4, HBGF-4Transforming protein KS3, heparin secretory transforming protein 1, Heparin secretory-transforming protein 1, Heparin-binding growth factor 4, HST-1, HST-1HSTF-1, HSTF1fibroblast growth factor 4 splice isoform, HSTFGF-4, human stomach cancer, transforming factor from FGF-related oncogene, kaposi sarcoma oncogene, KFGF, K-FGF, KS3, oncogene HST
Gene Symbols FGF4
Host Species Rat
Immunogen E. coli-derived recombinant mouse FGF-4 Ala30-Leu202 Accession # NP_034332
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2249
Test Specificity Detects mouse FGF-4 in direct ELISAs. In direct ELISAs, 10% - 50% cross-reactivity with recombinant mouse (rm) FGF-10, rmFGF-7, recombinant human (rh) FGF-16, rhFGF-20 and less than 5% cross-reactivity with rmFGF acidic, rmFGF basic, rmFGF-6, -8b, -8c, -15, -17, -21, -23, rhFGF acidic, rhFGF basic, rhFGF-3, -4, -5, -9, -11, -12, -13, -18, -19, or -22 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.