Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse FGFR2 (IIIc) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB716SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB716SP 25 μg
MAB716100 100 μg
MAB716500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB716SP Supplier R&D Systems Supplier No. MAB716SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 4 publications

FGFR2 Monoclonal specifically detects FGFR2 in Mouse samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen FGFR2
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 133706
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.1-0.4 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, FGF R2, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25
Gene Symbols FGFR2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse FGF R2 alpha (IIIc) Extracellular domains
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2263
Test Specificity Detects IIIc isoforms of mouse FGF R2 in direct ELISAs and Western blots. Does not react with IIIb isoforms of recombinant mouse (rm) FGF R2. In direct ELISAs and Western blots, 100% cross-reactivity with IIIc isoforms of recombinant human (rh) FGF R2 is observed, and no cross-reactivity with IIIb isoforms of rhFGF R2 or any isoforms of rhFGF R1, rmFGF R3, or rhFGF R4 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.