Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse FGFR4 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2265
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2265 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2265 Supplier R&D Systems Supplier No. BAF2265
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

FGFR4 Polyclonal specifically detects FGFR4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen FGFR4
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. CAA42551
Gene Alias CD334, CD334 antigen, EC 2.7.10, EC 2.7.10.1, FGF R4, FGFR-4, fibroblast growth factor receptor 4, JTK2hydroxyaryl-protein kinase, MGC20292, protein-tyrosine kinase, TKF, tyrosine kinase related to fibroblast growth factor receptor, tyrosylprotein kinase
Gene Symbols FGFR4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse FGF R4 Leu19-Asp366 Accession # CAA42551
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2264
Test Specificity Detects mouse FGF R4 in Western blots. In Western blots, approximately 35% cross-reactivity with recombinant human (rh) FGF R4 is observed and less than 5% cross-reactivity with rhFGF R1 alpha (IIIb), rhFGF R1 alpha (IIIc), rhFGF R1 beta (IIIb), rhFGF R1 beta (IIIc), recombinant mouse (rm) FGF R2 alpha (IIIb), rmFGF R2 alpha (IIIc), rmFGF R2 beta (IIIb), rmFGF R2 beta (IIIc), rmFGF R5, and rmFGF R5 beta is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.