Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse FGL2 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | FGL2/Fibroleukin |
---|---|
Clone | 829321 |
Dilution | Western Blot 2 μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB5257SP
![]() |
R&D Systems
MAB5257SP |
25 μg |
Each for $200.50
|
|
|||||
MAB5257100
![]() |
R&D Systems
MAB5257100 |
100 μg |
Each for $502.50
|
|
|||||
Description
FGL2/Fibroleukin Monoclonal specifically detects FGL2/Fibroleukin in Mouse samples. It is validated for Western Blot.Specifications
FGL2/Fibroleukin | |
Western Blot 2 μg/mL | |
Monoclonal | |
Lyophilized | |
Mouse | |
P12804 | |
10875 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
829321 | |
Western Blot | |
Unconjugated | |
Rat | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
fibrinogen-like 2, Fibrinogen-like protein 2, fibroleukin, pT49T49, T49 | |
Chinese Hamster Ovary cell line, CHO-derived mouse FGL2, Val20-Pro432, Accession # P12804 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse FGL2 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title