Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Fibroblast Activation Protein alpha/FAP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9727SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9727SP 25 μg
MAB9727100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9727SP Supplier R&D Systems Supplier No. MAB9727SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Fibroblast Activation Protein alpha/FAP Monoclonal specifically detects Fibroblast Activation Protein alpha/FAP in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry.

Specifications

Antigen Fibroblast Activation Protein alpha/FAP
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 983802
Conjugate Unconjugated
Dilution ELISA, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P97321
Gene Alias 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Fibroblast Activation Protein alpha/FAP Leu26-Asp761 Accession # P97321
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 2191
Test Specificity Detects human Fibroblast Activation Protein alpha/FAP in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.