Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse FIZZ1/RELM alpha Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
$200.50 - $549.50
Specifications
Antigen | FIZZ1/RELM alpha |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
FIZZ1/RELM alpha Polyclonal specifically detects FIZZ1/RELM alpha in Mouse samples. It is validated for Western Blot.Specifications
FIZZ1/RELM alpha | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
1810019L16Rik, Fizz1, Fizz-1, HIMF, RELM alpha, RELMa, RELMalpha, RELM-alpha, RETNLA, Retnla resistin like alpha, RETNLB, Xcp2 | |
RETNLA | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9EP95 | |
57262 | |
E. coli-derived recombinant mouse FIZZ1/RELM alpha Asp24-Ser111 Accession # Q9EP95 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse FIZZ1/RELM alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse Resistin is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title